missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, GlutaMAX™ Supplement
GREENER_CHOICE

Product Code. 11504456
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504456 500 mL 500mL
11514456 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11504456 Supplier Gibco™ Supplier No. 31980022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

IMDM, GlutaMAX™ Supplement

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. Gibco™ IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ IMDM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ IMDM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type Iscove's modified Dulbecco's medium
With Additives GlutaMAX, Phenol Red, Sodium Pyruvate
Without Additives No α±-Thioglycerol, No 2-Mercaptoethanol
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 6.9 to 7.3
Content And Storage 2 to 8°C
Description Iscove's Modified Dulbecco's Medium, 500mL, Liquid, 1 X Concentration, 6.9 to 7.3 pH, With Glutamax™, Phenol Red, Sodium Pyruvate, 12 Months Shelf Life, 270 to 310mOsm/kg Osmolality, For Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.