missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, powder

Product Code. 11514496 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 L
1 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11514496 10 L 10 L
10184932 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11514496 Supplier Gibco™ Supplier No. 42200030
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IMDM, powder

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ IMDM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ IMDM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Iscove's modified Dulbecco's medium
With Additives Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No α±-Thioglycerol, No 2-Mercaptoethanol
For Use With (Application) Mammalian cell culture
Quantity 10 L
Certifications/Compliance ISO 13485 standards, FDA
pH 4.4 to 4.8
Content And Storage 2 to 8°C
Description Iscove's Modified Dulbecco's Medium, 10L, Powder, 4.4 to 4.8 pH, With L-Glutamine, Phenol Red, Sodium Pyruvate, 36 Months Shelf Life, 215 to 245mOsm/kg Osmolality, For Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.