missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%

Product Code. 10088289 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10088289 1 g 1g
10230750 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10088289

Brand: Thermo Scientific Acros 432780010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64951-08-2 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64951-08-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD03419462
InChI Key WQLJLPDGSLZYEP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, h-imidazo 1,2-a pyridine-2-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic, 1h-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, technical grade
PubChem CID 2062168
IUPAC Name imidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C=CC=CC2=N1

Specifications

Melting Point 232.0°C to 234.0°C
Color White
Elemental Analysis 51.20 to 55.47% C, 4.30 to 4.65% H, 14.96 to 16.17% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Imidazo¢1,2-a!pyridine-2-carboxylic acid hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.