missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.5

Product Code. 15435719
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435719 100 mL 100mL
15445719 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435719 Supplier Thermo Scientific Chemicals Supplier No. J63496.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Imidazole
compliance-icons compliance-icons
Product Identifier
  • Imidazole, 0.5M buffer solution, pH 7.5
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Imidazole, Hydrochloric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.