Learn More
Imidazole-4-acetic acid monohydrochloride, 97%
Description
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 221°C to 224°C |
| Odor | Odorless |
| Quantity | 1 g |
| Beilstein | 3701591 |
| Solubility Information | Soluble in water. |
| Formula Weight | 162.58 |
| Percent Purity | 97% |
| Chemical Name or Material | Imidazole-4-acetic acid monohydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.