missing translation for 'onlineSavingsMsg'
Learn More

Imidocarb, TRC

Product Code. 30137770
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137770 100 g 100g
30132710 10 g 10g
30040835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137770 Supplier Toronto Research Chemicals Supplier No. TRCI387550100G
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27885-92-3
Molecular Formula C19 H20 N6 O
Molecular Weight (g/mol) 348.4
Synonym Carbanilide, 3,3'-di-2-imidazolin-2-yl- (8CI), N,N'-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea, 3,3'-Bis-(2-imidazolin-2-yl)carbanilide, Imidocarb, Piro-Stop
IUPAC Name 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea
SMILES O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4

Specifications

Quantity 100 g
InChI Formula InChI=1S/C19H20N6O/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26)
Formula Weight 348.1699
Chemical Name or Material Imidocarb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.