missing translation for 'onlineSavingsMsg'
Learn More

Iminodibenzyl 5-Carbonyl Chloride, TRC

Product Code. 30008116
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008116 100 mg 100mg
30136595 10 g 10g
30135359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008116 Supplier Toronto Research Chemicals Supplier No. TRCI408100100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33948-19-5
Molecular Formula C15 H12 Cl N O
Molecular Weight (g/mol) 257.71
Synonym 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carbonyl Chloride, 5-(Chlorocarbonyl)-10,11-dihydrodibenzazepine, 5-Chlorocarbonyl-10,11-dihydro-5H-dibenz[b,f]azepine, Carbamazepine Dihydro Chlorocarbonyl Impurity
IUPAC Name 5,6-dihydrobenzo[b][1]benzazepine-11-carbonyl chloride
SMILES ClC(=O)N1c2ccccc2CCc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H12ClNO/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-8H,9-10H2
Formula Weight 257.0607
Chemical Name or Material Iminodibenzyl 5-Carbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.