missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imipenem monohydrate, 98%

Product Code. 15595136 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595136 1 g 1g
15505146 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595136 Supplier Thermo Scientific Chemicals Supplier No. 455320010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Imipenem monohydrate,Tienam, Primaxin, CAS # 74431-23-5 is a broad-spectrum carbapenem antibiotic that interferes with the activity of penicillin binding proteins along the inner cell wall.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
• Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
• Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria

Applications
• Suitable for the study of antibiotic resistance
• High-quality, laboratory grade for use in vitro and in vivo experiments

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74431-23-5
Molecular Formula C12H19N3O5S
Molecular Weight (g/mol) 317.36
MDL Number MFCD09753321
InChI Key GSOSVVULSKVSLQ-XMCAPODCSA-N
PubChem CID 5282372
ChEBI CHEBI:51799
IUPAC Name (5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
SMILES O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O

Specifications

Type Imipenem Monohydrate
Melting Point 193.0°C to 198.0°C
Infrared Spectrum Authentic
Water 5 to 8%
Quantity 1 g
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material Imipenem monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.