missing translation for 'onlineSavingsMsg'
Learn More

Imipenem monohydrate, MedChemExpress

Product Code. 16428746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428746 Supplier Medchem Express Supplier No. HYB1369
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Imipenem monohydrate, a stable crystalline derivative of thienamycin, is an antibiotic and has the excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria. Imipenem monohydrate can be used for the research of carbapenem-nonsusceptible and P. aeruginosa biofilm infections.

Imipenem monohydrate, a stable crystalline derivative of thienamycin, is an antibiotic and has the excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria. Imipenem monohydrate can be used for the research of carbapenem-nonsusceptible and P. aeruginosa biofilm infections.

Specifications

Chemical Name or Material Imipenem monohydrate
Physical Form Powder
CAS 74431-23-5
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H19N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym N-Formimidoyl thienamycin monohydrateMK0787 monohydrate
Solubility Information H2O : 7.14 mg/mL (22.50 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(C(N12)=C(SCCNC=N)C[C@]2([H])[C@@H]([C@H](O)C)C1=O)O.O
Molecular Weight (g/mol) 317.36
Formula Weight 317.36
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.