missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Imiquimod, TLR7 Ligand

Product Code. 18606528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606528 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18606528 Supplier R&D Systems™ Supplier No. NBP2262281mg
  • ONLINE EXCLUSIVE PRICE
    229.18 €
    186.90 € / 1mg
    Save 42.28 €
    18% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Functional, In vitro assay, Ligand Activation

Imiquimod, TLR7 ligand is an imidazoquinoline amine analog to guanosine. As a synthetic molecule of the Imidazoquinoline family, it has potent immunostimulatory activity. Imiquimod has been shown to activate only TLR7, this activation is MyD88 dependant and leads to the induction of the transcription factor NF-kB.

  • Species: Human, Mouse
  • Specificity: Imiquimod is human/mouse TLR7 agonist
  • Stimulation of TLR7 can be achieved with 5-10μg/mL

Recommended Storage

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
For Use With (Application) For activation of TLR7
Product Line Novus
Product Type Imiquimod, TLR7 Ligand
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.