missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One IMP CT™ 96-well Protein Crystallization Plate

Product Code. 13024973
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Unit Size
13024973 Conical Bottom (V-well) Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13024973 Supplier Greiner Bio-One Supplier No. 673101
  • ONLINE EXCLUSIVE PRICE
    1285.00 €
    1173.43 € / Pack of 80
    Save 111.57 €
    8% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in microbatch crystallization. Greiner Bio-One IMP@CT™ 96-well Protein Crystallization Plate features conical wells that avoid spreading of droplets.

  • Low Profile
  • Polystyrene
  • Conical wells
  • Flat Bottom
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Product Type Crystallization Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Microbatch crystallization
Profile Low
Quantity 80/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.