missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Importin 9 Recombinant Rabbit Monoclonal Antibody (K01_1K59)
GREENER_CHOICE

Product Code. 30828453
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828453 100 μL
30830417 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828453 Supplier Invitrogen™ Supplier No. MA563169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Importin 9 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Functions in nuclear protein import as nuclear transport receptor. Serves as receptor for nuclear localization signals (NLS) in cargo substrates. Is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. The directionality of nuclear import is thought to be conferred by an asymmetric distribution of the GTP-and GDP-bound forms of Ran between the cytoplasm and nucleus. Mediates the nuclear import of H2B histone, RPS7 and RPL18A. Prevents the cytoplasmic aggregation of RPS7 and RPL18A by shielding exposed basic domains. May also import H2A, H3, H4 histones, RPL4 and RPL6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin 9
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1K59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene IPO9
Gene Accession No. Q96P70
Gene Alias 0710008K06Rik; AI845704; C78347; DKFZp761M1547; FLJ10402; HSPC273; Imp9; imp9a; imp9b; importin 9; importin-9; importin-9a; importin-9b; Ipo9; KIAA1192; mKIAA1192; Ran binding protein 9; ran-binding protein 9; RANBP9; RP11-90L20.4
Gene Symbols IPO9
Host Species Rabbit
Immunogen A synthetic peptide of human Importin 9.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55705
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.