Learn More
Importin alpha 3/KPNA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76300-0.025mg
 View more versions of this product
Product Code. 18536074
  • 189.00 € / 25µg
Description
Importin alpha 3/KPNA4 Polyclonal antibody specifically detects Importin alpha 3/KPNA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Importin alpha 3/KPNA4
Polyclonal
Unconjugated
PBS
importin alpha 3, Importin alpha Q1, importin subunit alpha-4, importin-alpha-Q1, IPOA3, karyopherin alpha 4 (importin alpha 3), MGC12217, MGC26703, Qip1, QIP1Karyopherin subunit alpha-4, SRP3
Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA4. The immunogen is located within amino acids 30 - 80 of KPNA4. Amino Acid Squence: RNVPQEDICEDS
0.025 mg
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL
EAW78632
Rabbit
Peptide affinity purified
RUO
3840
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.