missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INCENP Monoclonal Antibody (58-217)
GREENER_CHOICE

Product Code. 10332803
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10332803 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10332803 Supplier Invitrogen™ Supplier No. 392800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

INCENP Monoclonal Antibody for Western Blot, ICC/IF, IP, ELISA

Chromosomal passenger proteins are members of a group of proteins that move from centromeres to the spindle midzone during mitosis. INCENP (Inner Centromere Protein) is a chromosomal passenger protein with an essential role in mitosis and meiosis. It is a complex highly basic multidomain protein that is dynamically translocated from metaphase chromosomes to the spindle midzone during or just prior to anaphase. Some INCENP transfers to the equatorial cortex before formation of the cleavage furrow. Ultimately, it is discarded in the midbody at the completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INCENP
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 58-217
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Incenp
Gene Accession No. Q9NQS7
Gene Alias 2700067E22Rik; AU019509; binds and activates aurora-B and -C in vivo and in vitro; C130081E20; C77457; chromosomal passenger protein; Incenp; Inner centromere protein; inner centromere protein antigens 135/155kDa; inner centromere protein INCENP
Gene Symbols Incenp
Host Species Mouse
Immunogen Recombinant human INCENP (inner centromere protein).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3619
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.