missing translation for 'onlineSavingsMsg'
Learn More

Indane-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10704682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10704682 5 g 5g
10047939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10704682 Supplier Thermo Scientific Acros Supplier No. 437990050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52205-85-3
Molecular Formula C9H9ClO2S
Molecular Weight (g/mol) 216.679
InChI Key SWLIXMXSCZYVTQ-UHFFFAOYSA-N
Synonym indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
PubChem CID 3142583
IUPAC Name 2,3-dihydro-1H-indene-5-sulfonyl chloride
SMILES C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 45°C to 50°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 216.69
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Indane-5-sulfonyl chloride
compliance-icons
Product Identifier
  • Indane-5-sulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.