missing translation for 'onlineSavingsMsg'
Learn More

Indirubin-3'-monoxime, MedChemExpress

Product Code. 16483857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483857 50 mg 50mg
16457918 10 mg 10mg
16400388 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483857 Supplier Medchem Express Supplier No. HY19807
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Indirubin-3'-monoxime is a potent GSK-3β inhibitor, and weakly inhibits 5-Lipoxygenase, with IC50s of 22 nM and 7.8-10 μM, respectively; Indirubin-3'-monoxime also shows inhibitory activities against CDK5/p25 and CDK1/cyclin B, with IC50s of 100 and 180 nM.

Indirubin-3'-monoxime is a potent GSK-3β inhibitor, and weakly inhibits 5-Lipoxygenase, with IC50s of 22 nM and 7.8-10 μM, respectively

Specifications

Chemical Name or Material Indirubin-3'-monoxime
Physical Form Solid
CAS 160807-49-8
Color Black
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H11N3O2
For Use With (Application) Cancer-Kinase/protease
Synonym Indirubin-3'-oxime
Solubility Information DMSO : ≥ 37 mg/mL (133.44 mM)
SMILES O=C1NC2=C(C=CC=C2)/C1=C3NC4=C(C=CC=C4)C\3=N/O
Molecular Weight (g/mol) 277.28
Formula Weight 277.28
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.