missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indisulam, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that targets the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of the activation of both CDK2 and cyclin E. Indisulam (E 7070) targets splicing by inducing RBM39 degradation via recruitment to DCAF15.
Specifications
Specifications
| Chemical Name or Material | Indisulam |
| CAS | 165668-41-7 |
| Color | Light Nude |
| Purity Grade Notes | Research |
| Molecular Formula | C14H12ClN3O4S2 |
| Quantity | 50 mg |
| Synonym | E 7070 |
| Solubility Information | DMSO : 100 mg/mL (259.17 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC=C(S(=O)(N)=O)C=C1)(NC2=CC=CC3=C2NC=C3Cl)=O |
| Molecular Weight (g/mol) | 385.85 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction