missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldoxime, 97%, Thermo Scientific™

Product Code. 15460958
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460958 5 g 5g
15470958 250 mg 250mg
15450958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460958 Supplier Thermo Scientific Alfa Aesar Supplier No. H54754.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2592-05-4
MDL Number MFCD00022737

Specifications

Molecular Formula C9H8N2O
Quantity 5 g
Synonym 1h-indole-3-carbaldehyde oxime, 1h-indole-3-carboxaldehyde, oxime, e-1h-indole-3-carbaldehyde oxime, hydroxyimino indol-3-ylmethane, indole oxime, e-n-hydroxy-1-1h-indol-3-yl methanimine, indole-3-aldoxime, n-1h-indol-3-ylmethylidene hydroxylamine, e-n-1h-indol-3-ylmethylidene hydroxylamine, indole-3-aldehyde oxime
InChI Key HFNHXOHLZXORGW-VOTSOKGWSA-N
SMILES C1=CC=C2C(=C1)C(=CNO)C=N2
IUPAC Name N-[(Z)-indol-3-ylidenemethyl]hydroxylamine
Molecular Weight (g/mol) 160.18
PubChem CID 5310834
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Indole-3-carboxaldoxime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.