missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Infigratinib phosphate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Infigratinib phosphate (BGJ-398 phosphate
Specifications
Specifications
| Chemical Name or Material | Infigratinib phosphate |
| CAS | 1310746-10-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H34Cl2N7O7P |
| Quantity | 10 mg |
| Synonym | BGJ-398 phosphateNVP-BGJ398 phosphate |
| SMILES | O=P(O)(O)O.ClC1=C(C(Cl)=C(C=C1OC)OC)NC(N(C2=CC(NC3=CC=C(C=C3)N4CCN(CC4)CC)=NC=N2)C)=O |
| Molecular Weight (g/mol) | 658.47 |
| Formula Weight | 658.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction