missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Antibody - (A/California/14/2009), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30320SS
This item is not returnable.
View return policy
Description
Influenza A H1N1 Polyclonal antibody specifically detects Influenza A H1N1 in Virus samples. It is validated for Western Blot.
Specifications
| Influenza A H1N1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HA | |
| A/California/14/2009 (H1N1) virions were used as an immunogen. | |
| 0.025 mg | |
| Primary | |
| This antibody detects the following H1N1 proteins: Hemagglutinin (HA) ∼75 kDa, Neuraminidase monomer (NA) ∼55 kDa, Matrix (M) ∼26 kDa and Non-structural Protein monomer (NS) ∼26-27 kDa. Reacts with A/New Caledonia/20/99, A/Brisbane/10/2007, A/California/14/2009 and likely reacts with most other isolates. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000 | |
| Flu | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 956529 | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction