Learn More
Invitrogen™ Influenza A H1N1 HA (A/Puerto Rico/8/34) Recombinant Rabbit Monoclonal Antibody (016)

Rabbit Recombinant Monoclonal Antibody
159.00 € - 429.00 €
Specifications
| Antigen | Influenza A H1N1 HA (A/Puerto Rico/8/34) |
|---|---|
| Clone | 016 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30533935
![]() |
Invitrogen™
MA560326 |
100 μL |
429.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535902
|
Invitrogen™
MA560327 |
20 μL |
159.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recognizes Influenza H1N1 (A/Puerto Rico/8/1934) Hemagglutinin/HA; No cross-reactivity in ELISA with: H1N1 (A/Victoria/4897/2022) HA Protein; H1N1 (A/Wisconsin/588/2019)/(A/Victoria/2570/2019) HA Protein; H1N1 (A/Wisconsin/67/2022) HA Protein; H3N2 (A/Darwin/6/2021) HA Protein; H3N2 (A/Darwin/9/2021) HA Protein; Influenza B (B/Austria/1359417/2021) HA Protein; Influenza B (B/PHUKET/3073/2013) HA Protein; Influenza B (B/Washington/02/2019) HA Protein.
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.Specifications
| Influenza A H1N1 HA (A/Puerto Rico/8/34) | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Recombinant Influenza H1N1 (A/Puerto Rico/8/1934) Hemagglutinin/HA protein (Met1-Gln528). | |
| Antibody |
| 016 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.