Learn More
Invitrogen™ Influenza A H3N2 HA (A/Brisbane/10/2007) Recombinant Rabbit Monoclonal Antibody (014), Biotin
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560308
 View more versions of this product
Product Code. 30534442
  • 569.00 € / 100µg
Description
Influenza A H3N2 HA (A/Brisbane/10/2007) Recombinant Monoclonal Antibody for ELISA
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A H3N2 HA (A/Brisbane/10/2007)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA
014
Biotin
flu hemagglutinin; H3N2 HA; H3N2 hemagglutinin; Influenza H3N2; influenza hemagglutinin
Recombinant Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin/HA protein (Met1-Ile531).
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles