missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPR1 Monoclonal Antibody (377029)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561491

Product Code. 30533750

  • 459.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Guanylyl cyclases, catalyzing the production of cGMP from GTP, are classified as soluble and membrane forms. The membrane guanylyl cyclases, often termed guanylyl cyclases A through F, form a family of cell-surface receptors with a similar topographic structure: an extracellular ligand-binding domain, a single membrane-spanning domain, and an intracellular region that contains a protein kinase-like domain and a cyclase catalytic domain. GC-A and GC-B function as receptors for natriuretic peptides; they are also referred to as atrial natriuretic peptide receptor A and type B. Also see NPR3, which encodes a protein with only the ligand-binding transmembrane and 37-amino acid cytoplasmic domains. NPR1 is a membrane-bound guanylate cyclase that serves as the receptor for both atrial and brain natriuretic peptides and BNP, respectively).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPR1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
P16066
NPR1
Mouse myeloma cell line NS0-derived recombinant human NPRA/NPR1, Gly33-Glu473.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
377029
Unconjugated
NPR1
AI893888; ANPa; ANP-A; ANPRA; ANPR-A; atrial natriuretic peptide A-type receptor; atrial natriuretic peptide receptor 1; atrial natriuretic peptide receptor A; atrial natriuretic peptide receptor type A; atrionatriuretic peptide receptor A; Gca; GC-A; Guanylate cyclase A; guanylyl cyclase-A; GUC2A; GUCY2A; natriuretic peptide A type receptor; natriuretic peptide receptor 1; natriuretic peptide receptor A; natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptide receptor A); Nppa receptor; Npr1; Npra; NPR-A; Pndr; testicular tissue protein Li 20
Mouse
Protein A/G
RUO
4881
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.