Learn More
Invitrogen™ Influenza A H7N9 HA Recombinant Rabbit Monoclonal Antibody (001)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560312
Description
Has cross-reactivity in ELISA with: H7N9 (A/Anhui/1/2013) HA Protein; H7N9 (A/Hangzhou/1/2013) HA Protein; H7N9 (A/Hangzhou/3/2013) HA Protein; H7N9 (A/Pigeon/Shanghai/S1069/2013) HA Protein; H7N9 (A/Shanghai/1/2013) HA Protein; H7N9 (A/Shanghai/2/2013) HA Protein; H7N9 (A/Shanghai/4664T/2013) HA Protein; H7N9 (A/Zhejiang/1/2013) HA Protein; H7N9 (A/Zhejiang/DTID-ZJU10/2013) HA Protein; H7N7 (A/chicken/Netherlands/1/03) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N2 (A/ruddy turnstone/New Jersey/563/2006) HA Protein; H7N3 (A/turkey/Italy/214845/2002) HA Protein; H7N8 (A/mallard/Netherlands/33/2006) HA Protein; H10N3 (A/duck/Hong Kong/786/1979) HA Protein; H15N8 (A/duck/AUS/341/1983) HA Protein; No cross-reactivity in ELISA with: H1N1 (A/California/07/2009) HA; H1N1 (A/California/04/2009) HA0 Protein; H2N2 (A/Canada/720/2005) HA Protein; H3N2 (A/Brisbane/10/2007) HA Protein; H4N6 (A/mallard/Ohio/657/2002) HA Protein; H5N1 (A/Anhui/1/2005) HA Protein; H6N1 (A/northern shoveler/California/HKWF115/2007) HA Protein; H7N1 (A/turkey/Italy/4602/99) HA1 Protein; H7N3 (A/chicken/SK/HR-00011/2007) HA1 Protein; H7N7 (A/equine/Kentucky/1a/1975) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N7 (A/Netherlands/219/2003) HA1 Protein; H8N4 (A/pintail duck/Alberta/114/1979) HA Protein; H9N2 (A/Hong Kong/1073/99) HA Protein; H11N2 (A/duck/Yangzhou/906/2002) HA Protein; H12N5 (A/green-winged teal/ALB/199/1991) HA Protein; H13N8 (A/black-headed gull/Netherlands/1/00) HA Pr...
Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.
Specifications
| Influenza A H7N9 HA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA | |
| 001 | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| Recombinant Influenza A H7N9 HA protein. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.