Learn More
Invitrogen™ Influenza A H7N9 HA Recombinant Rabbit Monoclonal Antibody (002), Biotin
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560310
 View more versions of this product
Product Code. 30533736
  • 569.00 € / 100µg
Description
Influenza A H7N9 HA Recombinant Monoclonal Antibody for ELISA
Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A H7N9 HA
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA
002
Biotin
flu hemagglutinin; influenza hemagglutinin
Recombinant Influenza A H7N9 HA protein.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles