missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A NP Recombinant Rabbit Monoclonal Antibody (014), Biotin, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552355
This item is not returnable.
View return policy
Description
The nucleoprotein (NP) of Influenza A virus encapsidates the negative strand of the viral RNA and is essential for replicative transcription. It may also be involved in other essential functions throughout the virus life cycle. As well as binding ssRNA, NP is able to self associate to form large oligomeric complexes. NP is able to interact with a variety of other macromolecules of both viral and cellular origins. It binds the PB1 and PB2 subunits of the polymerase and the matrix protein M1. NP has also been shown to interact with at least four cellular polypeptide families: nuclear import receptors of the importin class, filamentous (F) actin, the nuclear export receptor CRM1 and a DEAD box helicase BAT1/UAP56 (Portela et al 2002).
Specifications
| Influenza A NP | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA | |
| 014 | |
| Biotin | |
| Influenza nucleoprotein | |
| Recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein/NP Protein (Met1-Asn498). | |
| 20 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction