missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza B NP Recombinant Rabbit Monoclonal Antibody (036)

Product Code. 30536187
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30536187 100 μg 100µg
30535492 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30536187 Supplier Invitrogen™ Supplier No. MA560373
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Widely recognize Influenza B Nucleoprotein. Has cross-reactivity in ELISA and WB with: Influenza B (B/Austria/1359417/2021) NP Protein; Influenza B (B/Brisbane/60/2008) NP; Influenza B (B/Colorado/06/2017) NP; Influenza B (B/Florida/4/2006) NP; Influenza B (B/Phuket/3073/2013) NP; Influenza B (B/Washington/02/2019) NP. Has cross-reactivity in ELISA with: Influenza B (B/Victoria/705/2018) NP; No cross-reactivity in ELISA and WB with: H1N1 (A/Brevig Mission/1/1918) NP; H1N1 (A/Brisbane/02/2018 NP; H1N1 (A/California/07/2009) NP; H1N1 (A/Guangdong-Maonan/SWL1536/2019) NP; H1N1 (A/Hawaii/70/2019) NP; H1N1 (A/Michigan/45/2015 NP; H1N1 (A/Puerto Rico/8/34/Mount Sinai) NP (I116M); H1N1 (A/Victoria/2570/2019)/(A/Wisconsin/588/2019) NP; H2N2 (A/Ann Arbor/6/1960) NP; H3N2 (A/Aichi/2/1968) NP; H3N2 (A/Cambodia/e0826360/2020 (H3N2)-like NP; H3N2 (A/Hong Kong/1/1968) NP; H3N2 (A/Hong Kong/2671/2019) NP; H3N2 (A/Hong Kong/45/2019) NP; H3N2 (A/Hong Kong/4801/2014) NP; H3N2 (A/Kansas/14/2017) NP; H3N2 (A/Switzerland/9715293/2013) NP; H5N1 (A/Texas/37/2024) NP Protein; H7N9 (A/Anhui/1-BALF_RG6/2013) NP; H7N9 (A/Shanghai/2/2013) NP; No cross-reactivity in ELISA with: H1N1 (A/Victoria/2454/2019) NP; SARS-CoV-2 Nucleocapsid Protein; HCoV-229E Nucleocapsid Protein HCoV-NL63 Nucleocapsid Protein; HCoV-HKU1 Nucleocapsid Protein; HCoV-OC43 Nucleocapsid Protein; No cross-reactivity in WB with: H3N2 (A/Darwin/9/2021)/(A/Darwin/6/2021) NP Protein.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B NP
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 036
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Influenza B; Influenza NP
Host Species Rabbit
Immunogen Recombinant Influenza B (B/Florida/4/2006) Nucleoprotein/NP protein (Met1-Tyr560).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.