missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza H14N5 Hemagglutinin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
299.00 € - 507.00 €
Specifications
| Antigen | Influenza H14N5 Hemagglutinin |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637997
|
Novus Biologicals
NBP3-12739-100ul |
100 μg |
507.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639966
|
Novus Biologicals
NBP3-12739-50ul |
50 μg |
299.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza H14N5 Hemagglutinin Polyclonal antibody specifically detects Influenza H14N5 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza H14N5 Hemagglutinin | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Produced in rabbits immunized with purified, recombinant Influenza H14N5 Hemagglutinin (Uniprot#: P26136; Met1-Lys342). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title