missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza H18N11 Hemagglutinin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12750-50ul
This item is not returnable.
View return policy
Description
Influenza H18N11 Hemagglutinin Polyclonal antibody specifically detects Influenza H18N11 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza H18N11 Hemagglutinin | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Influenza H18N11 Hemagglutinin (Accession#: AGX84934.1; Met1-Arg339). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction