missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ING1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15604445
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15604445 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604445 Supplier Invitrogen™ Supplier No. PA550511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ING1 protein.

This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ING1
Applications Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ING1
Gene Accession No. Q9QXV3, Q9UK53
Gene Alias 2610028J21Rik; AA407184; AI875420; growth inhibitor ING1; growth inhibitory protein ING1; ING1; inhibitor of growth family member 1; inhibitor of growth family, member 1; inhibitor of growth protein 1; mING1h; p24ING1c; p33; p33ING; p33ING1; p33ING1b; p33ING1c; p37Ing1b; p47; p47ING1a; tumor suppressor ING1
Gene Symbols ING1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human inhibitor of growth family, member 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26356, 3621
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.