missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Antibody (06), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06215-100ul
This item is not returnable.
View return policy
Description
ING5 Monoclonal antibody specifically detects ING5 in Human samples. It is validated for ELISA
Specifications
| ING5 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| ELISA 1:1000-1:2000 | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ING5 (Uniprot#: Q8WYH8-2; Glu36-Gln84). | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction