missing translation for 'onlineSavingsMsg'
Learn More

INH6, MedChemExpress

Product Code. 16449636
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449636 50 mg 50mg
16423078 10 mg 10mg
16404376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16449636 Supplier Medchem Express Supplier No. HY100541
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress INH6 is a potent Nek2/Hec1 inhibitor; inhibits the growth of HeLa cells with an IC50 of 2.4 μM.

INH6 is a potent Nek2/Hec1 inhibitor

Specifications

Chemical Name or Material INH6
Physical Form Solid
CAS 1001753-24-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H18N2OS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (155.08 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C2=C(C)C=C(C)C=C2C)=CS1)C3=CC=CC=C3
Molecular Weight (g/mol) 322.42
Formula Weight 322.42
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.