Learn More
Invitrogen™ INHBB Recombinant Rabbit Monoclonal Antibody (23GB6625), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553080
 View more versions of this product
Product Code. 30089419
  • 446.00 € / 100µL
Description
The inhibin beta B subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. Because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. Furthermore, the beta B subunit forms a homodimer, activin B, and also joins with the beta A subunit to form a heterodimer, activin AB, both of which stimulate FSH secretion.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
INHBB
Recombinant Monoclonal
2.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09529, P17491, Q04999
INHBB
A synthesized peptide derived from human Inhibin beta B (350-407AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB6625
Unconjugated
INHBB
activin AB beta polypeptide; activin betaB; activin beta-B chain; INHBB; Inhibin beta B chain; inhibin beta B subunit; inhibin beta subunit B; inhibin beta-B; inhibin, beta B; Inhibin, beta-2
Rabbit
Affinity chromatography
RUO
16324, 25196, 3625
-20°C