missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin alpha Antibody (SR1673), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22103-100ul
This item is not returnable.
View return policy
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Inhibin alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1673 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200 | |
| A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
| A synthesized peptide derived from human Inhibin alpha (Uniprot #: P05111) | |
| 100 μg | |
| Apoptosis, Cancer | |
| 3623 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction