missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Inhibin beta A Antibody (SR4080), Novus Biologicals™
Rabbit Monoclonal Antibody
339.00 € - 406.00 €
Specifications
| Antigen | Inhibin beta A |
|---|---|
| Clone | SR4080 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041750
|
Novus Biologicals™
NBP3-27818-50ul |
50 μL |
339.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041755
|
Novus Biologicals™
NBP3-27818-100ul |
100 μL |
406.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibin beta A Monoclonal antibody specifically detects Inhibin beta A in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| Inhibin beta A | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 3624 | |
| IgG | |
| Affinity purified |
| SR4080 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1 | |
| A synthesized peptide derived from human Inhibin beta A (Uniprot #: P08476) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title