missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INMT Antibody (S08-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19663-100ul
This item is not returnable.
View return policy
Description
INMT Monoclonal antibody specifically detects INMT in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| INMT | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-9C6 | |
| Unconjugated | |
| Amine N-methyltransferase, Aromatic alkylamine N-methyltransferase, Arylamine N-methyltransferase, EC 2.1.1, EC 2.1.1.49, Indolamine N-methyltransferase, indolethylamine N-methyltransferase, MGC125940, MGC125941, nicotine N-methyltransferase | |
| Recombinant protein of human INMT (Uniprot # O95050) | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 11185 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction