missing translation for 'onlineSavingsMsg'
Learn More

INS Mouse anti-Human, CoraLite 488, Clone: 4B6A7, Proteintech

Product Code. p-7227559 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17711289 20 μL 20µL
16830746 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17711289 Supplier Proteintech Supplier No. CL4886619820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The INS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INS. This antibody recognizes human, mouse, rat antigen. The INS antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.

Specifications

Antigen INS
Applications Immunofluorescence
Classification Monoclonal
Clone 4B6A7
Conjugate CoraLite 488
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Rat, Mouse
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.