missing translation for 'onlineSavingsMsg'
Learn More

inS3-54A18, MedChemExpress

Product Code. 16424856
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424856

Brand: Medchem Express HY103128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress inS3-54A18 is a potent STAT3 inhibitor, with anti-cancer properties.

inS3-54A18 is a potent STAT3 inhibitor, with anti-cancer properties.

Specifications

Chemical Name or Material inS3-54A18
CAS 328998-53-4
Color Orange
Purity Grade Notes Research
Molecular Formula C23H16ClNO2
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (401.25 mM)
SMILES O=C1N(C2=CC=C(O)C=C2)C(C3=CC=CC=C3)=C/C1=C/C4=CC=C(Cl)C=C4
Molecular Weight (g/mol) 373.83
Formula Weight 373.83
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.