missing translation for 'onlineSavingsMsg'
Learn More

INSL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

386.00 € - 540.00 €

Specifications

Antigen INSL3
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18426060
View Documents
Novus Biologicals
NBP1-81223-25ul
25ul
386.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18288235
View Documents
Novus Biologicals
NBP1-81223
0.1 mL
540.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INSL3 Polyclonal specifically detects INSL3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

INSL3
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
insulin-like 3, insulin-like 3 (Leydig cell), leydig insulin -like hormone, leydig insulin -like peptide, Leydig insulin-like peptide, Ley-I-L, MGC119818, MGC119819, Relaxin-like factor, relaxin-like factor b, RLFley-I-L, RLNL
INSL3
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Polyclonal
Rabbit
Human
P51460, P51460, P51460, P51460
3640
This antibody was developed against Recombinant Protein corresponding to amino acids:EMREKLCGHHFVRALVRVCGGPRWSTEARRPATGGDRELLQWLERRHLLHGLVADSNLTLGPGLQPLPQTSHHHRHHRAAATNPARYCCLSGCTQQDLLTLCPY
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.