missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSL6 Monoclonal Antibody (421401)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561437
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene contains a classical signature of the insulin superfamily and is significantly similar to relaxin and relaxin-like factor. This gene is preferentially expressed in testis. Its expression in testis is restricted to interstitial cells surrounding seminiferous tubules, which suggests a role in sperm development and fertilization.
Specifications
| INSL6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y581 | |
| INSL6 | |
| E. coli-derived recombinant human INSL6, Arg21-Tyr213. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 421401 | |
| Unconjugated | |
| INSL6 | |
| INSL6; insulin like 6; insulin-like 6; insulin-like peptide 5; insulin-like peptide 6; Insulin-like peptide INSL6; Insulin-like peptide INSL6 A chain; Insulin-like peptide INSL6 B chain; Relaxin/insulin-like factor 1; RIF1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 11172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur