Learn More
Invitrogen™ INSR beta Recombinant Rabbit Monoclonal Antibody (HL3132)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556763
Product Code. 30276074
  • 500.00 € / 100µL
Description
INSR beta Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
INSR beta
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P06213, P15208
INSR
Recombinant fragment of human Insulin Receptor beta.
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL3132
Unconjugated
INSR
4932439J01Rik; CD220; CD220 beta; CD221; D630014A15Rik; HHF5; IGF-I receptor; Insr; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR beta; IR-A; IR-B
Rabbit
Protein A
RUO
16337, 3643
-20°C, Avoid Freeze/Thaw Cycles