missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ INSTA-Blot Mouse Tissues

Product Code. p-200085296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Membrane
2 Membrane
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18097054 1 Membrane Each
18455252 2 Membrane Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18097054 Supplier R&D Systems™ Supplier No. NBP230111
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: SDS-Page

  • Screening proteins from various mouse tissues
  • Contains 11 mouse tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
  • Prepared from resolution by SDS-PAGE at 125V for 1 hour
  • Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
  • Stained with amido black for visualization of proteins

Applications:

Screening proteins from various mouse tissues
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-Page
Quantity 1 Membrane
Material PVDF
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.