missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2b/CD41, Rabbit anti-Human, Clone: 7E3 (Abciximab), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81051-0.025MG
This item is not returnable.
View return policy
Description
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry, Block/Neutralize.
Specifications
| Integrin alpha 2b/CD41 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Block/Neutralize | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises human Integrin alpha 2b/CD41. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Flow Cytometry, Blocking Assay | |
| 7E3 (Abciximab) | |
| Unconjugated | |
| PBS | |
| ITGA2B | |
| CD41 is the integrin alpha chain 2b forming a fibrinogen receptor in heterodimeric complex with beta 3. | |
| 0.025 mg | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction