Learn More
Integrin alpha 5/CD49e Antibody (CL6940) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76518MFV610
Product Code. 30296494
  • 780.00 € / 0.10mL
Description
Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 5/CD49e
Monoclonal
mFluor Violet 610 SE
CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
This antibody was developed using a recombinant protein derived from P08648, with the exact immunogen sequence remaining proprietary.
0.1 mL
CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells
3678
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL6940
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified