missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5/CD49e Antibody (CL6945) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76519G
This item is not returnable.
View return policy
Description
Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Integrin alpha 5/CD49e | |
| Monoclonal | |
| DyLight 488 | |
| CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5 | |
| This antibody was developed using a recombinant protein derived from P08648, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells | |
| 3678 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL6945 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction