missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha L/CD11a Antibody (MHM24) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
262.00 € - 641.00 €
Specifications
| Antigen | Integrin alpha L/CD11a |
|---|---|
| Clone | MHM24 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681308
|
Novus Biologicals
NBP3-09043-0.025mg |
0.025 mg |
262.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642647
|
Novus Biologicals
NBP3-09043-0.2mg |
0.2 mg |
641.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha L/CD11a Monoclonal antibody specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| Integrin alpha L/CD11a | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
| Spleen cells from BALB/c mice immunised with EBV-transformed human lymphoblastoid cells were fused with P3-NSUlAg4.1 myeloma cells and the resulting hybridomas were screened for antibody production by a radioimmunoassay on T and B cell lines. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MHM24 | |
| Flow Cytometry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 3683 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title