missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (1B4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
262.00 € - 615.00 €
Specifications
| Antigen | Integrin beta 2/CD18 |
|---|---|
| Clone | 1B4 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638437
|
Novus Biologicals
NBP3-09040-0.2mg |
0.2 mg |
615.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632389
|
Novus Biologicals
NBP3-09040-0.025mg |
0.025 mg |
262.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human, Macaque samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune AssaysSpecifications
| Integrin beta 2/CD18 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Macaque | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Blood was obtained from 4 helathy donors and the mononuclear cells harvested and grown. A BALB/c mouse was injected with 10^6 monocytes three times at monthly intervals, and four times with 5x10^6 monocytes oon each of the four days before fusion. The spleen cells were then fused to P3U1 myeloma cells to generate hybridomas and RIA was used to identify lines of interest. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1B4 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cytokine Research, Immunology | |
| PBS | |
| 3689 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title