missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (10B10D5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
217.00 € - 543.00 €
Specifications
| Antigen | Integrin beta 4/CD104 |
|---|---|
| Clone | 10B10D5 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610786
|
Novus Biologicals
NBP2-37392-0.025ml |
0.025 mL |
217.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047785
|
Novus Biologicals
NBP2-37392 |
0.1 mL | |||||||
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Integrin beta 4/CD104 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| ITGB4 | |
| IgG1 | |
| Unpurified |
| 10B10D5 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| Ascites with 0.03% Sodium Azide | |
| 3691 | |
| Purified recombinant fragment of human ITGB4 (AA: 1619-1822) expressed in E. Coli. | |
| Primary | |
| 202 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title