missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (308601), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4054A
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 308601 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 4/CD104, Asn29-Ser711, Accession # NP_001005608 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3691 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction