missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (308601), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4054CL1
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 4/CD104, Asn29-Ser711, Accession # NP_001005608 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| 308601 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction